Learn More
AdipoGen Palmarumycin C3
SDP
Supplier:  AdipoGen BVT0078M005
Encompass_Preferred
Chemical. CAS 159934-11-9. Formula C20H12O6. MW 348.3. Isolated from Sphaeropsidales sp. Rasfarnesyltransferase inhibitor. Antifungal, antibacterial and herbicidal. Structurally unique natural products. The basic structure of palmarumycin C1 can be modified by a number of hydroxylation, oxygenation, dehydrogenation and chlorination steps. Antioxidant
Catalog No. 50-256-2116
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.