Learn More
Cayman Chemical Palmitic Acid, 57-10-3, 25 g
SDP
Supplier:  Cayman Chemical 1000662725
Encompass_Preferred
Molecular Formula C16H32O2, Purity >98%, Formula Weight 256.4, Synonyms: hexadecanoic acid; A common 16-carbon saturated fatty acid that represents 10-20% of the normal human dietary fat intake.
Catalog No. NC1155404
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.