Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Palmitic acid, 98%

Catalog No. AC129700250
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC129700250 25 kg Fibre drum
AC129702500 250 g Plastic bottle
AC129700010 1 kg Plastic bottle
AC129700025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC129700250 Supplier Thermo Scientific Chemicals Supplier No. 129700250
Add to Cart

CAS: 57-10-3 | C16H32O2 | 256.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-10-3
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00002747
InChI Key IPCSVZSSVZVIGE-UHFFFAOYSA-N
Synonym palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
PubChem CID 985
ChEBI CHEBI:15756
IUPAC Name hexadecanoic acid
SMILES CCCCCCCCCCCCCCCC(O)=O

Specifications

Acidity 215 to 233mg KOH/g
Melting Point 59.0°C to 63.0°C
Color White
Density 0.8500g/mL
Boiling Point 351.5°C
Flash Point 206°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Fibre drum
Linear Formula CH3(CH2)14CO2H
Quantity 25 kg
Saponification Value 216 to 224mg KOH/g
Beilstein 02,IV,1157
Merck Index 15,7097
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot alcohol, ether and propyl alcohol
Viscosity 7.8 mPa.s (70°C)
Formula Weight 256.42
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material Palmitic acid
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-312-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.