Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitic Acid, Approx. 92%, Spectrum™ Chemical
SDP

Catalog No. 18612116 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-116 12 kg Poly Pail
18-612-119 500 g Poly Pail
18-612-117 2.5 kg Poly Pail
18-612-118 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-116 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PA11012KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57-10-3
Molecular Weight (g/mol) 256.42 g/mol

Specifications

CAS Max % 1
Melting Point 59° to 64°C
Boiling Point 350°C
Packaging Poly Pail
Quantity 12 kg
Percent Purity ∼92%
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material Palmitic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.