Learn More
Palmitic Acid, FCC, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 350°C |
| Ignition Residue | 0.001 |
| Melting Range | 61°C to 64°C |
| Packaging | Poly Bottle |
| Quantity | 2.5 kg |
| Saponification Value | 205-221 |
| Grade | FCC |
| Physical Form | Crystals or Flakes |
| Chemical Name or Material | Palmitic Acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.