Learn More
Bachem Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH
SDP
Supplier:  Bachem 4015000.0250
Encompass_Preferred
Palmitoyl-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH, 87420-41-5, C54H103NO7S, Mr 910.48, 250mg. Pam3Cys-OH. The lipophilic cysteine derivative Pam3Cys resembles the N-terminus of the lipoprotein from E.coli outer membranes. Pam3Cys has been successfully used in the synthesis of immunogenic conjugates like peptide mitogens or vaccines. For coupling protocols see the papers of Zeng et al.
Catalog No. 50-259-178
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.