Learn More
Bachem Palmitoyl-Gly-Gln-Pro-Arg-OH
SDP
Supplier:  Bachem 4145516.2004
Encompass_Preferred
Pal-GQPR or Palmitoyl Tetrapeptide-7/3 is a synthetic peptide that is used in cosmetics to suppress the production of excess interleukins, the chemical messengers which trigger the body’s acute inflammatory response. Therefore Pal-GQPR can inhibit unnecessary inappropriate inflammatory responses and glycation damage.
Catalog No. NC2404495
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.