Learn More
Abcam (±)-Palmitoylcarnitine Chloride, Protein Kinase C Inhibitor, Solid, 100 mg
Description
- Activity: Protein kinase C (PKC) inhibitor
- Molecular Weight: 436.1 Da
- Purity: >98%
- Physical State: Solid
- Biological Function: Intermediate in long-chain fatty acid metabolism; capable of disrupting cell membranes
- Storage Temperature: -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Protein Kinase C |
| Substrate Properties | Lipid-based Substrate |
| Buffer | 100% (±)-Palmitoylcarnitine chloride |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% (±)-Palmitoylcarnitine chloride |
| Quantity | 100 mg |
| For Use With (Application) | Protein kinase C (PKC) inhibition, Fatty acid metabolism research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.