Learn More
Selleck Chemical LLC Palomid 529 (P529) 10mg 914913-88-5 SG 00529
SDP
Supplier:  Selleck Chemical LLC S223810MG
Encompass_Preferred
Palomid 529 (P529, SG 00529) inhibits both the mTORC1 and mTORC2 complexes, reduces phosphorylation of pAktS473, pGSK3?S9, and pS6. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-645-1
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.