Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort PAM™ 100% Dual Layer Memory Foam Insole

Ergonomically designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Max shock absorption

$35.40

Specifications

Color Black, Blue, Yellow
For Use With (Application) Stationary, mobile and working outdoors
For Use With (Equipment) Industrial, safety and steel toe footwear
Material Memory Foam
Gender Unisex
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-813-493
View Documents
MEGAComfort
PAMW567
Women's: 5-7
Pair of 1 for $35.40
Only null left
Add to Cart
 
Description

Description

  • Personal Anti-Fatigue Mat (PAM™) insole
  • Uses patented combination of high density dual layer 100% memory foam
  • Reduces pain and fatigue while increasing balance and comfort
  • For industrial, safety and steel toe footwear
  • Ideal for stationary, mobile and working outdoors
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti-perspiration and anti-odor
  • Temperature regulating aeration holes
  • Machine washable
Specifications

Specifications

Black, Blue, Yellow
Industrial, safety and steel toe footwear
Unisex
PAM Insole
Stationary, mobile and working outdoors
Memory Foam
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.