Learn More
Lkt Laboratories CAS 109552-15-0, ≥98%, It is used to prevent injury- or chemotherapy-induced bone loss. It increases muscle fiber, bone strength, and bone density.
SDP
Supplier:  Lkt Laboratories P0049
Encompass_Preferred
Pamidronate is a bisphosphonate that exhibits anti-resorptive and anti-osteoporotic activities. Pamidronate is clinically used to prevent injury- or chemotherapy-induced bone loss. Clinically, pamidronate increases muscle fiber, bone strength, and bone density. This compound also prevents cancer metastasis to bone tissue.
Catalog No. NC2527363
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.