Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pamoic Acid Disodium Salt Monohydrate 98.0+%, TCI America™
SDP

Catalog No. P1059250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1059250G 250 g
P105925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1059250G Supplier TCI America Supplier No. P1059250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6640-22-8
Molecular Formula C23H14Na2O6
Molecular Weight (g/mol) 432.339
MDL Number MFCD00036183
InChI Key YGLLICRFEVEWOZ-UHFFFAOYSA-L
Synonym sodium pamoate, pamoic acid disodium salt, disodium pamoate, unii-rpo7lte47e, disodium 4,4'-methylenebis 3-hydroxy-2-naphthoate, rpo7lte47e, disodium methylenebis 2-hydroxy-3-naphthoate, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy-, disodium salt, sodium 4,4'-methylenebis 3-hydroxy-2-naphthoate, embonic acid disodium salt
PubChem CID 54676156
IUPAC Name disodium;3-carboxy-1-[(3-carboxy-2-oxidonaphthalen-1-yl)methyl]naphthalen-2-olate
SMILES C1=CC=C2C(=C1)C=C(C(=C2CC3=C(C(=CC4=CC=CC=C43)C(=O)O)[O-])[O-])C(=O)O.[Na+].[Na+]

Specifications

Color Yellow
Quantity 250 g
Formula Weight 432.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pamoic Acid Disodium Salt Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.