Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PAMP-20 (human)
GREENER_CHOICESDP

Catalog No. 65521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-521 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 65-521 Supplier Tocris Bioscience™ Supplier No. 6552/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

MRGPRX2 agonist

PAMP-20 (human) is an endogenous peptide agonist of Mas related GPR X2 (MRGPRX2, EC50 = 251 nM). Also a noncompetitive nicotinic cholinergic antagonist. Inhibits nicotine-stimulated catecholamine secretion from PC12 cells in vitro, and from sympathetic nerve endings and adrenal chromaffin cells in vivo. Corresponds to amino acids 1 - 20 of proadrenomedullin. Antihypertensive.

Specifications

Purity 95%
CAS 150238-87-2
Content And Storage Store at -20°C
Molecular Weight (g/mol) 2460.87
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.