Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAMR1 Mouse anti-Human, Clone: OTI4E2, lyophilized, TrueMAB™
SDP

Catalog No. 501669394
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-9394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-9394 Supplier OriGene Supplier No. CF809285
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

PAMR1 may be involved in regeneration of skeletal muscle. Although related to peptidase S1 family, lacks the conserved active Ser residue in position 665 which is replaced by a Thr, suggesting that it has no protease activity.

Specifications

Antigen PAMR1
Applications Western Blot
Classification Monoclonal
Clone OTI4E2
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PAMR1
Gene Accession No. Q6UXH9
Gene Alias DKFZP586H2123, FP938, RAMP
Gene Symbols PAMR1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 338-559 of human PAMR1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25891
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.