Learn More
METTLER TOLEDO Pan 100 μL Al
Catalog No. 01674716 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Product Type:
Crucible
Catalog No. Product Type
01-674-716 Crucible
1 options
Catalog No. 01-674-716 Supplier METTLER TOLEDO Supplier No. 51143092
Add to Cart
Add to Cart
Set of 100 crucibles without lids. This large crucible is used for DSC measurements. It allows you to use larger quantities of sample if the signal obtained from the sample in a 40 μL pan is too weak. Crucible presses sold separately.
  • Made of durable aluminum
  • 100 μL volume for larger DSC sample quantities
  • Withstands temperatures up to 640°C
  • Operates at 0 MPa pressure
  • No lids and no pin included
  • Simple and easy to use
  • Compatible with METTLER TOLEDO crucible presses (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Capacity (Metric) 100 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder