Learn More
METTLER TOLEDO Pan 40 μL Cu
Catalog No. 01674717 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Material:
Copper
Catalog No. Material
01-674-717 Copper
1 options
Catalog No. 01-674-717 Supplier METTLER TOLEDO Supplier No. 51140407
Add to Cart
Add to Cart
Set of 100 crucibles without lids, primarily used for determining oxidative stability (OIT). The induction time measured is typically compared with results obtained using an inert aluminum pan. Compatible with crucible presses sold separately.
  • Made of durable copper for reliable performance
  • 40 μL volume suitable for DSC measurements
  • Withstands temperatures up to 750°C
  • Operates at 0 MPa pressure
  • No lids and no pin included
  • Supplied as a set of 100 crucibles
  • Designed for precise and consistent thermal analysis
TRUSTED_SUSTAINABILITY
Specifications
Material Copper
Capacity (Metric) 40 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder