Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan Acetylation Mouse anti-Canine, Human, Mouse, Rat, Clone: 1D5F2, Proteintech
SDP

Catalog No. p-7213461 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6721 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6721 Supplier Proteintech Group Inc Supplier No. 662891IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is used to detect the Acetylation of Lysine. The ∽17 kDa in our detection is Acetylated Histone.

Acetylation and De-acetylation are the important reaction in epigenetic. Many proteins, especially Histone, are with Acetylated of Lysine.

Specifications

Antigen Pan Acetylation
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1D5F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Alias Acetylation, Acetylation Lysine, Pan Acetylation
Host Species Mouse
Immunogen Peptide
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.