Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pan B Cells Monoclonal Antibody (BB6-10A10), FITC
GREENER_CHOICE

Catalog No. PIMA528749
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528749 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528749 Supplier Invitrogen™ Supplier No. MA528749
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pan B Cells Monoclonal Antibody for Flow

Pan B Cells antibody specific anti equine reagents have yet to be fully characterized for the typically recognized B-cell makers such as CD19, CD20, CD21, CD22 and CD79. While testing has demonstrated the cross reactivity of some monoclonal B-cell markers raised against other species with equine B cells, such as the Mouse anti Human CD79a antibody, (clone HM57), an overview of which may be found in the report of the second equine leucocyte antigen workshop. Pan B Cells antibody is directed against equine Ig light chains. Characterization studies demonstrated that clone CVS36 bound 100% of CD5-ve peripheral blood lymphocytes and recognized all equine B-cells (Lunn et al. 1998) As the antigen recognized by clone CVS36 appears to be present on the surface of all equine B-cells it is therefore is a reagent that can be used as a pan B-cell marker for domestic horses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pan B Cells
Applications Flow Cytometry
Classification Monoclonal
Clone BB6-10A10
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with <0.1% sodium azide; pH 7.4
Host Species Mouse
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.