Learn More
R&D Systems™ Pan Caspase OPH Inhibitor Q-VD, Non-omethylated
SDP
Catalog No. OPH00101M Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
OPH00101M 1 mg
1 options
This item is not returnable. View return policy
Catalog No. OPH00101M Supplier R&D Systems™ Supplier No. OPH00101M
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Cell-permeable, irreversible inhibitor of caspase activity. Q-VD-OPh does not exhibit toxic effects at typical working concentrations.
Members of the caspase gene family (cysteine proteases with aspartate specificity) play significant roles in both inflammation and apoptosis.
  • The IC50 range for caspases -1, -3, -8, -9, -10, and -12 is 25 - 400 nM
  • Cells that are induced to undergo apoptosis can be cultured in the presence or absence of Q-VD-OPh inhibitor
  • The Caspase Inhibitor binds to the active site of activated proteases
  • Treated cells can then be assayed for evidence of apoptosis inhibition by examining either whole cells or cell lysates using standard apoptosis assays
  • More effective in preventing apoptosis than the widely used pan Caspase Inhibitor, ZVAD-fmk
Specifications
Product Type Inhibitor
Formula Weight 513.77 d
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 mg