Learn More
Biocare Medical, LLC Pan Cytokeratin [AE1/AE3] Antibody - Predilute, 25 ml
SDP
Supplier:  Biocare Medical, LLC PM011H
Encompass_Preferred
Pan Cytokeratin [AE1/AE3] antibody recognizes the acidic and basic (Type I and II) subfamilies of cytokeratins. The cocktail of these two antibodies has been shown to detect in human epithelia. The acidic cytokeratins have molecular weights of 56.5, 55, 51, 50, 50, 48 46, 45 and 40 kDa. The basic cytokeratins have molecular weights of 65-67, 64, 59, 58, 56 and 52 kDa. In immunohistochemistry studies, this Pan Cytokeratin antibody has proven useful as a screener for the majority of human carcinomas.
Catalog No. 50-830-03
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.