Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan-keratin Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735329 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5329 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5329 Supplier Proteintech Group Inc Supplier No. 264111AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is a pan-keratin antibody.

The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.

Specifications

Antigen pan-keratin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KRT10
Gene Accession No. P02533, P02538, P04264, P05787, P08727, P08729, P08779, P12035, P13645, P13647, P19012, P19013, P35908
Gene Alias 58 kDa cytokeratin, CK 5, CK5, Cytokeratin 5, K5, keratin 5, KRT5, KRT5A, Type II keratin Kb5
Gene Symbols KRT1, KRT10, KRT14, KRT15, KRT16, KRT19, KRT2, KRT3, KRT4, KRT5, KRT6A, KRT7, KRT8
Host Species Rabbit
Immunogen KRT5 Fusion Protein Ag24184
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3848, 3849, 3850, 3851, 3852, 3853, 3855, 3856, 3858, 3861, 3866, 3868, 3880
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.