Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan-PAX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733397 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3397 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3397 Supplier Proteintech Group Inc Supplier No. 213831AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody is a Pan-PAX antibody.

This gene encodes a member of the paired box (PAX) family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.

Specifications

Antigen Pan-PAX
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PAX1
Gene Accession No. O43316, O88436, P09084, P15863, P23759, P23760, P24610, P26367, P32114, P32115, P47239, P47242, P51974, P55771, P63015, P63016, Q00288, Q02548, Q02650, Q02962, Q06710, Q2L4T2
Gene Alias paired box, paired box 8, Paired box protein Pax, Paired box protein Pax 8, Pan-PAX, PAX 8, PAX8
Gene Symbols Pax1, Pax2, PAX3, PAX4, PAX5, PAX6, PAX7, Pax8, PAX9
Host Species Rabbit
Immunogen Pan-PAX Fusion Protein Ag16018
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114502, 18503, 18504, 18505, 18506, 18507, 18508, 18509, 18510, 18511, 25509, 293992, 311505, 362741, 500453, 500574, 5075, 5076, 5077, 5078, 5079, 5080, 5081, 5083, 7849, 81819, 83630
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.