Learn More
ABclonal Technology Pan Phospho-Tyrosine
SDP
Supplier:  ABclonal Technology AP1316100UL
Encompass_Preferred
Tyrosine phosphorylation (pTyr) much of which occurred on localized multiple sites initiates cellular signaling governs cellular functions and its dysregulation is implicated in many diseases especially cancers pTyr-specific sensing is of great significance for understanding disease states and developing targeted anticancer drugs
Catalog No. 50-000-13947
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.