Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18604959 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-959 1 g Amber Glass Bottle
18-604-960 5 g Amber Glass Bottle
18-604-961 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-959 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P10001GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1000, 85-85-8, C15H11N3O

Spectrum™ Chemical PAN, Reagent, ACS

Chemical Identifiers

CAS 85-85-8
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.27
InChI Key RAXUMGMWXZYADR-SDXDJHTJSA-N
IUPAC Name (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 g
Grade Reagent
Physical Form Crystals or Flakes
Chemical Name or Material PAN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.