Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Pancell T: Serum-Free, Chemically-Defined T-Cell Expansion Medium (10 x 500 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04710414X10
Encompass_Preferred
PAN-Biotech Pancell T: Serum-Free, Chemically-Defined T-Cell Expansion Medium (10 x 500 ml). BASAL MEDIUM ONLY. DESCRIPTION: PAN-Biotech Pancell T is a chemically defined and protein-free (< 0.02%) medium for serum-free cultivation and proliferation of lymphocytes from full blood. The medium can be used for the enrichment and expansion of T-cells in vitro. Pancell T enables: 1) Enrichment of T-cells from PBMC / full blood, after stimulation (e.g., with PHA-L or anti-CD3), 2) Stable performance > 4 passages, 3) Long-term cultivation of immortalized T-cells, 4) CD3+ > 80%. CELL TYPE: T-/CIK/ L AK Cells. CULTURE TYPE: Suspension Culture. COMPOSITION: Pancell T: T-Cell Expansion Medium (Cat. No. P04-710414X10, P04-710414X20): consists of a balanced mixture of salts, amino acids, vitamins, trace elements and hormones for optimized growth.
Catalog No. 50-238-8558
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.