Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Pancell T: Serum-Free, Chemically-Defined T-Cell Expansion Kit (20 x 500 ml kit)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04710414KX20
Encompass_Preferred
PAN-Biotech Pancell T: Serum-Free, Chemically-Defined T-Cell Expansion Kit (20 x 500 ml kits). DESCRIPTION: PAN-Biotech Pancell T is a chemically defined and protein-free (< 0.02%) medium for serum-free cultivation and proliferation of lymphocytes from full blood. The medium can be used for the enrichment and expansion of T-cells in vitro. Pancell T enables: 1) Enrichment of T-cells from PBMC / full blood, after stimulation (e.g., with PHA-L or anti-CD3), 2) Stable performance > 4 passages, 3) Long-term cultivation of immortalized T-cells, 4) CD3+ > 80%. CELL TYPE: T-/CIK/ L AK Cells. CULTURE TYPE: Suspension Culture COMPOSITION: Pancell T: T-Cell Expansion Medium consists of a balanced mixture of salts, amino acids, vitamins, trace elements and hormones for optimized growth. In addition to Pancell T basal medium, this kit includes PHA-L as supplement for the stimulation of lymphocytes (Cat. No. P05-01010M) and a cytokines supplement (Cat. No. P04-413S).
Catalog No. 50-238-8634
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.