Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Pancoll Pro, Human Blood Cell Separation Medium, Low Endotoxin (<0,1 EU/ml), 20 x 100 ml
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0460105X20
Encompass_Preferred
PAN-Biotech Pancoll Pro, Human Blood Cell Separation Medium, Low Endotoxin (<0.1 EU/ml), 20 x 100 ml. DESCRIPTION: In many cases the isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. Separating solutions are used for this purpose. Pancoll separating solutions from PAN-Biotech contain a polysaccharide with a molecular weight of 400,000 daltons. This hydrophilic polymer allows production of aqueous solutions for cell separation with a density of up to 1.119 g/ml. PAN-Biotech offers a variety of ready-to-use products with a density of 1.063 g/ml up to 1.119 g/ml for a wide range of cell separation applications. DENSITY: 1.100 g/ml. ENDOTOXIN: Less than 0.1 EU/ml LPS. Without EDTA. Without HEPES. STERILE: yes. STORAGE: +2°C - RT. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-238-8578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.