Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pancreatic Amylase Monoclonal Antibody (AMY10-102.5)
GREENER_CHOICE

Catalog No. 01671571
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-571 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-571 Supplier Invitrogen™ Supplier No. MA110618
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10618 detects Amylase from human samples. MA1-10618 has been successfully used in ELISA applications. The MA1-10618 immunogen is purified human pancreatic Amylase.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch andglycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pancreatic Amylase
Applications ELISA
Classification Monoclonal
Clone AMY10-102.5
Concentration ~1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene AMY2B
Gene Accession No. P04746, P19961
Gene Alias 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Symbols AMY2A, AMY2B
Host Species Mouse
Immunogen Purified human pancreatic Amylase.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279, 280
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.