Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic Lipase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985917
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5917 Supplier Bioss Supplier No. BS4125R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the lipase gene family. It encodes a carboxyl esterase that hydrolyzes insoluble, emulsified triglycerides, and is essential for the efficient digestion of dietary fats. This gene is expressed specifically in the pancreas.

Specifications

Antigen Pancreatic Lipase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PNLIP
Gene Accession No. P16233
Gene Alias 1810007A24Rik; pancreatic lipase; pancreatic lipase-related protein 2; Pancreatic triacylglycerol lipase; pancreatic triglyceride lipase; PANLI; PL; Pnlip; PNLIPD; PNLIPRP2; PTL; RP11-33D13.1; triglyceride lipase
Gene Symbols PNLIP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Pancreatic Lipase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5406
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.