Learn More
Diagnostic Biosystems PANCREATIC POLYPEPTIDE Pancreatic Polypeptide
Supplier:  Diagnostic Biosystems RP03005
Encompass_Preferred
This antibody reacts with pancreatic polypeptide. It stains the periphery of islets, exocrine pancreatic parenchyma and the epithelium of small and medium sized ducts and acinar cells.
Catalog No. 50-238-5442
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.