Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ PANCREATIN 4X from Porcine Pancreas

Catalog No. MP219397520 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219397520 20 g
MP219397580 100 g
MP219397590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219397520 Supplier MP Biomedicals™ Supplier No. 0219397520
Add to Cart
Add to Cart

Used to treat conditions in which pancreatic secretions are deficient, such as surgical pancreatectomy, pancreatitis and cystic fibrosis.

Applications: Pancreatin is used to treat conditions in which pancreatic secretions are deficient, such as surgical pancreatectomy, pancreatitis and cystic fibrosis. It has been claimed to help with food allergies, celiac disease, autoimmune disease, cancer and weight loss.
Product Description: Pancreatin is a mixture of digestive enzymes produced by the exocrine cells of the pancreas. It is composed of amylase, lipase and protease. The trypsin found in pancreatin works to hydrolyze proteins into oligopeptides; amylase hydrolyzes starches into oligosaccharides and the disaccharide maltose; and lipase hydrolyzes triglycerides into fatty acids and glycerols.
Biochem/physiol Actions: Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40 °C, will digest not less than 25 times its weight of casein in 60 minutes at pH 7.5 at 40 °C and will release not less than 2 microequivalents of acid per min per mg pancreatin from olive oil at pH 9.0 at 37 °C.
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
For Use With (Application) Convert starch into soluble carbohydrates
Content And Storage 0°C
Quantity 20 g
Product Type Proteins, Enzymes & Peptides
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.