Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pancreatin, USP from Procine Pancreas

Catalog No. MP210255720 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210255720 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210255720 Supplier MP Biomedicals™ Supplier No. 0210255720
Add to Cart
Add to Cart

Hydrolyzes proteins, starch and fats

Pancreatin is a digestive enzyme produced by the pancreas and delivered to the small intestine for the hydrolysis of complex nutrients. Pancreatin contains enzymatic components including trypsin, amylase and lipase, ribonuclease, and protease, produced by the exocrine cells of the porcine pancreas.

TRUSTED_SUSTAINABILITY

Specifications

Color Off-white to tan
Grade USP
For Use With (Application) Cell Biology
Content And Storage -20°C
Source Hog pancreas
Quantity 20 g
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.