Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancuronium bromide, MP Biomedicals™

Catalog No. ICN15605380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
0.25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15605380 100 mg
ICN15605310 10 mg
ICN15605350 50 mg
ICN15605383 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15605380 Supplier MP Biomedicals, Inc Supplier No. 0215605380
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15500-66-0
Molecular Formula C35H62Br2N2O4+2
Molecular Weight (g/mol) 734.699
MDL Number MFCD00079223
InChI Key NPIJXCQZLFKBMV-YTGGZNJNSA-N
Synonym 1,1'-[2β,3α,5α,16b,17b-3-17-bis(acetyloxy)-androstane-2,16-diyl]bis[1-methylpiperidinium]dibromide
PubChem CID 129893486
IUPAC Name [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-acetyloxy-10,13-dimethyl-2,16-bis(1-methylpiperidin-1-ium-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;dihydrobromide
SMILES CC(=O)OC1CC2CCC3C(C2(CC1[N+]4(CCCCC4)C)C)CCC5(C3CC(C5OC(=O)C)[N+]6(CCCCC6)C)C.Br.Br

Specifications

Color White
Quantity 100 mg
Formula Weight 732.7
Physical Form Crystalline Powder
Chemical Name or Material Pancuronium bromide
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.