Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panexin BMM Serum Substitute with Defined Components, w/o: Glucocorticoide (100 ml)
Supplier: ILEX LIFE SCIENCES LLC P04951SA2
Panexin BMM Serum Substitute with Defined Components, WITHOUT: Glucocorticoid. QUANTITY: 100 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: -20 degrees Celsius. DESCRIPTION: Panexin BMM is a fully defined serum replacement for the cultivation of macrophages from mouse bone marrow (murine bone marrow derived macrophages, BMM) under serum-free conditions. The ready-to-use sterile solution in a final concentration of 5% is added to the basal medium RPMI 1640, supplemented with 50 micromolar Mercaptoethanol and 2 ng/ml GM-CSF murine recombinant. COMPOSITION: Panexin BMM contains purified proteins (less than 3% w/v), lipids, salts, amino acids, trace elements, attachment factors, hormones and animal-derived components (<12% w/v). It contains no growth factors, undefined hydrolysates or lysates (e. g. Peptones).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.