Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panexin BMM Serum Substitute with Defined Components for cultivation of macrophages from mouse bone marrow, w/o: Glucocorticoid (20 x 10 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04951SA2MX20
Encompass_Preferred
PAN-Biotech GmbH (Germany). INTRO: Panexin BMM is a fully defined serum replacement for cultivation of macrophages from mouse bone marrow (murine bone marrow derived macrophages, BMM) under serum-free conditions. The ready-to-use sterile solution in a final concentration of 5% is added to basal medium RPMI 1640, supplemented with 50 uM Mercaptoethanol and 2 ng/ml GM-CSF murine recombinant. As undefined components from serum-containing cultures are eliminated, results are more reproducible. In Panexin BMM, matured macrophages will show excellent attachment capabilities. COMPOSITION: Panexin BMM contains purified proteins (<3%), lipids, salts, amino acids, trace elements, attachment factors, hormones and animal-derived components (<12%). It contains no growth factors, undefined hydrolysates or lysates (e. g. Peptones). STORAGE: -20°C, in the dark.
Catalog No. NC3233507
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.