Learn More
ILEX LIFE SCIENCES LLC TC-100 INSECT MEDIUM 6 X 500ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0492500X6
Encompass_Preferred
PAN-Biotech TC-100 Insect Medium, w: L-Glutamine, w: 0.35 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 0.35 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: Without Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Dr. Gardiner and Dr. Stockdale created TC-100 Insect Medium by modifying the original version of Grace's insect medium. TC-100 is optimized for the growth of insect cells, including Spodeptera frugiperda (Sf9), and for the expression of recombinant Autographa californica Nuclear Polyhedrosis Virus (rAcNPV) using the Baculovirus Expression Vector System (BEVS). TC-100 is suitable for the growth of cells from a variety of lepidopteran species.
Catalog No. 50-273-3754
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.