Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panexin NTS, Serum Substitute with Defined Components for Suspension Cells (500 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0495850
Encompass_Preferred
Panexin NTS, Serum Substitute with Defined Components for Non-adherent (Suspension) Cells. QUANTITY: 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). STORAGE: -20 degrees Celsius. DESCRIPTION: PAN-Biotech Panexin NTS is a ready-to-use, fully defined serum substitute for the cultivation of suspension cells under serum-free conditions. Panexin NTS is developed with a unique technology and contains a special 3-dimensional substance release system (3D-SRS) for an optimal support of cells with nutrients and growth stimulants. The sterile solution is added to the culture medium in a final concentration of 10%. It supports the growth of many cell types in an optimum manner. COMPOSITION: Contains purified proteins, lipids, salts, amino acids, trace elements, and hormones in an optimized formulation and a new 3-dimensional substance release system (3D-SRS). It contains no growth factors, undefined hydrolysates or peptones. Animal-derived components are < 2% w/v.
Catalog No. 50-238-8610
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.