Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PANK3 Monoclonal Antibody (3D7)
GREENER_CHOICE

Catalog No. PIMA522583 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522583 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522583 Supplier Invitrogen™ Supplier No. MA522583
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MKIKDAKKPS FPWFGMDIGG TLVKLSYFEP IDITAEEEQE EVESLKSIRK YLTSNVAYGS TGIRDVHLEL KDLTLFGRRG NLHFIRFPTQ DLPTFIQMGR DKNFSTLQTV LCATGGGAYK FEKDFRTIGN LHLHKLDELD CLVKGLLYID SVSFNGQAEC YYFANASEPE RCQKMPFNLD DPYPLLVVNI GSGVSILAVH SKDNYKRVTG TSLGGGTFLG LCSLLTGCES FEEALEMASK GDSTQADKLV RDIYGGDYER FGLPGWAVAS SFGNMIYKEK RESVSKEDLA RATLVTITNN IGSVARMCAV NEKINRVVFV GNFLRVNTLS MKLLAYALDY WSKGQLKALF LEHEGYFGAV GALLGLPNFS.

This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is expressed most abundantly in the liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PANK3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PANK3
Gene Accession No. Q9H999
Gene Alias hPanK3; mPanK3; Pank3; Pantothenate kinase 3; pantothenic acid kinase 3
Gene Symbols PANK3
Host Species Mouse
Immunogen PANK3 (AAH13705, 1 a.a. ~ 370 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79646
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.