Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pannexin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 433270
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-327-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-327-0 Supplier Invitrogen™ Supplier No. 433270
  • $479.00
    $359.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Pannexin 3 Polyclonal Antibody for Western Blot, ICC/IF

Pannexins constitute a new family of gap junction type proteins. They are highly conserved in worms, mollusks, insects and mammals and are predicted to have four transmembrane regions, two extracellular loops, one intracellular loop and intracellular N- and C-termini. Besides the connexin proteins, Pannexin 1 and 2 seem to be molecular components of neuronal gap junctions ('electrical synapses') that are thought to form connections between principal cells in the hippocampus. Functional expression in paired Xenopus oocytes indicated that pannexins are capable of forming communicating junctions. In addition to forming gap junction channels in paired oocytes, Pannexin 1 can also form a mechanosensitive and ATP-permeable channel in the nonjunctional plasma membrane. Pannexin 1 and 3 are both glycoproteins with a cell surface distribution profile and life cycle dynamics distinct from connexin 43. Pannexin 3 expression was prevalent in skin and cartilage. In transiently expressing cells both pannexin 1 and 3 were incapable of forming intercellular channels but assembled into functional cell surface channels. Suggested roles in the ischemic death of neurons, schizophrenia, inflammation and tumor suppression have drawn attention for properties and cellular functions of pannexins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pannexin 3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PANX3
Gene Accession No. P60572, Q8CEG0, Q96QZ0
Gene Alias 3230401P04; 4833413G11Rik; gap junction protein pannexin 3; pannexin 3; pannexin-3; Panx3; Px3; si:dkey-263o8.2
Gene Symbols PANX3
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the mouse and rat Pannexin 3 protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116337, 208098, 315567
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.