Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Panobinostat
GREENER_CHOICE

Catalog No. p-7250129
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-291-0 10 mg
76-295-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 76-291-0 Supplier Tocris Bioscience™ Supplier No. 7629/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pan-histone deacetylase inhibitor

Panobinostat is a potent pan-histone deacetylase (HDAC) inhibitor (IC50 values = 2.1 - 531 nM). Panobinostat induces histone H3 and H4 acetylation and potently inhibits cell proliferation and cell viability in HH, BT474 and HCT116 cells (IC50 are 1.8, 2.6 and 7.1 nM, respectively) in vitro. It leads to significant tumor regression of up to 94% in an HH CTCL mouse xenograft model. Panobinostat inhibits the DNA binding activity of STAT5 in leukemia cell lines and acts synergistically with 17-AAG (Cat. No. 1515) to induce apoptosis. The compound shows potent antimyeloma activity, including in drug-resistant cell lines. Panobinostat also induces a significant increase in cell-associated unspliced (CA-US) HIV-1 RNA from CD4+ T cells and leads to activation of transcription of proviruses. Panobinostat simultaneously suppresses the expression of angiotensin-converting enzyme 2 (ACE2) and ABO in epithelial cell lines. Antimalarial and orally bioavailable.

Specifications

Chemical Name or Material (2 E)-N-Hydroxy-3-[4-[[[2-(2-methyl-1 H-indol-3-yl)ethyl]amino]methyl]phenyl]-2-propenamide
CAS 404950-80-7
Quantity 10 mg
Target Histone Deacetylase Inhibitors
Molecular Formula C21H23N3O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.