Learn More
AdipoGen PANoptosome h Starter Set
SDP
Supplier:  AdipoGen AG44B0012KI01
Encompass_Preferred
The PANoptosome Human Antibodies Starter Set is an all-in-one solution to study the three key modes of programmed cell death, including pyroptosis, apoptosis and necroptosis using Western blotting application. This antibody starter set comprises our KO extract validated STANDARD antibodies against the key components of the NLRP3 inflammasome including NLRP3, Asc and Caspase-1, the cytosolic RNA/DNA sensor ZBP1 and the apoptosis target caspase-8. These antibodies are used and published by the experts in inflammasome and cell death research. This economic starter set contains enough primary antibodies to perform at least 3 western blot experiments. Each antibody is available in 100µg catalog sizes for further experiments.
Catalog No. 50-256-2747
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.