Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panserin 401, Serum-free Allround Cell Culture Medium, w: L-Glutamine (6 x 500 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P04710401X6
Encompass_Preferred
Panserin 401, Serum-free Allround Cell Culture Medium, w: L-Glutamine. QUANTITY: 6 x 500 ml. MANUFACTURED BY: Pan-Biotech (Germany). DESCRIPTION: Panserin 401 is a fully defined ready-to-use medium for the serum-free cultivation of a multitude of adherent and non adherent (suspension) cells. Based on Iscove's MEM, trace elements, albumin, cholesterol, soy lipids and vitamins were added to the medium. It does not contain any growth or attachment factors. Protein and animal-derived components < 0.2 % w/v. Hydrolysates < 0.001 % w/v. APPLICATION: Panserin 401 is a multi-purpose medium suitable for a variety of cells. As the medium contains no growth factors there is a possibility to investigate the effects of specific growth factors added to the cell culture. Among others, Panserin 401 has been used successfully with: Hybridoma, Lymphocytes, Macrophages, Fibroblasts, Melanocytes, Carcinoma cells, HEK-cells, HeLa-cells, CHO-cells, Megakaryocytic cell lines (HEL, Dami, CMK), HL60-cells.
Catalog No. 50-238-8517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.