Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Panserin 413, Serum-free Medium for Lymphocytes, T-Cells and Hybridoma Cells, w: L-Glutamine, modified (20 x 1000 ml)
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0471413SX20
Encompass_Preferred
PAN-Biotech Panserin 413: Serum-Free Medium for Lymphocytes, T Cells, and Hybridoma Cells, w: L-Glutamine, modified. QUANTITY: 20 x 1000 ml. MANUFACTURED BY: Pan-Biotech (Germany). DESCRIPTION: Panserin 413 is a ready-to-use, serum-free, low protein (<0.1% w/v) and low animal-derived components (< 0.6%) medium for the serum-free cultivation and proliferation of lymphocytes from whole blood. The medium can be used for the enrichment of T cells , cytokine-induced killer (CIK) cells, lymphokine-activated killer (LAK) cells, and natural killers (NK) cells. COMPOSITION: This defined formulation based on RPMI 1640 and DMEM/F12 is optimized with additional salts, trace elements, albumin, cholesterol, lipids and vitamins. The medium contains no hydrolysates, and only traces of animal-derived components (< 0.6% w/v) and proteins (< 0.1% w/v).
Catalog No. 50-238-8482
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.