Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pantoprazole Sodium, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604973 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-973 1 g Amber Glass Bottle
18-604-974 5 g Amber Glass Bottle
18-604-975 25 g Amber Glass Bottle
18-604-976 100 g Amber Glass Bottle
18-604-977 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-973 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P37141GM
Add to Cart
Add to Cart

P3714, 164579-32-2, C16H14F2N3NaO4S·1.5H2O

Spectrum™ Chemical Pantoprazole Sodium, USP is used for short-term treatment of erosive esophagitis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 164579-32-2
Molecular Formula C16H18F2N3NaO5S
Molecular Weight (g/mol) 425.38
InChI Key QHNXIKDJFYGZTL-UHFFFAOYNA-N
IUPAC Name sodium 5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methanesulfinyl]-2,3-dihydro-1H-1,3-benzodiazol-1-ide hydrate
SMILES O.[Na+].COC1=CC=NC(CS(=O)C2NC3=CC(OC(F)F)=CC=C3[N-]2)=C1OC

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.