Learn More
Paoli Clay Company Dissection Pans and Pads FSE
SCI_ED
Catalog No. S17424
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
12.5 x 9 in.
Material:
Vinyl
Product Type:
Replacement deluxe Flex-pad
Catalog No. Dimensions (L x W) Material Product Type
S17424 12.5 x 9 in. Vinyl Replacement deluxe Flex-pad
1 options
This item is not returnable. View return policy
Catalog No. S17424 Supplier Paoli Clay Company Supplier No. ADPAD
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Keep specimens looking fresh between dissection periods.
  • High-density polyethylene (HDPE) pans offer tight-sealing lids for durability and easy cleanup
  • Each is available with Flex-Pads — odorless vinyl pads that are machine washable and reusable
  • Flex-Pads replace wax for faster setup and cleanup
Specifications
Product Type Replacement deluxe Flex-pad
Material Vinyl
For Use With (Application) Dissecting
Dimensions (L x W) 12.5 x 9 in.