Learn More
Specifications
Specifications
| Content And Storage | Room Temperature |
| Product Line | Novus |
| Quantity | 1 Each |
| For Use With (Application) | Preparation of Working Solutions 1. Ready to be Used Directly on Wet Slides. Protocol Recommendations 1. Hold The Pen Point up and Press it with Your Fingertip to Take Blocked Air Out From The Pen Barrel to Prevent Overflow. 2. To Start, Press The Tip of The Pen Straight Down Until Fluid Starts Soaking The Tip and Then Draw a Circle Around The Specimen. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
