Learn More
Abcam PAPA NONOate, Nitric o x ide (NO) donor, 5MG
Supplier:  Abcam AB1451965MG
Encompass_Preferred
MW 176.22 Da, Purity >97%. Nitric oxide (NO) donor. Has an NO release half life of 77 minutes. Regulates caspase-3 activity and suppresses caspase-mediated apoptosis. Shows anti-ischemic effects in vivo. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5406