Description
- EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
- EA-65 is used with OG-6 for non-gynecological staining.
- Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Specifications
Specifications
| Quantity | 2.5 gal. |
| Product Type | EA-65 Papanicolaou Stain |