Learn More
Epredia™ Papanicolaou OG-6 and Modified OG Stains
GSA_VA
Catalog No. 22050208 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1 gal.
2.5 gal.
Product Type:
Modified OG Stain
OG-6 Papanicolaou Stain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-208 1 gal. OG-6 Papanicolaou Stain
22-050-207 1 pt. OG-6 Papanicolaou Stain
22-050-305 2.5 gal. OG-6 Papanicolaou Stain
22-050-287 1 gal. Modified OG Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-050-208 Supplier Epredia™ Supplier No. 75211
Add to Cart
Add to Cart
This item is not returnable. View return policy
Stain gynecological specimens with Epredia™ Papanicolaou Stain OGs.
  • OG-6 stains the keratinized areas of the cell
  • Modified OG is a Gill modification that minimizes precipitation and reduces staining time
  • Use OG-6 with EA-36 or EA-50 when staining gynecological specimens
  • Use modified OG with EA-65 for staining non-gynecological specimens
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 1 gal.
Product Type OG-6 Papanicolaou Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov