Description
- Ready to use
- Staining solution for cytological cancer cycle diagnosis
- Used for cytological investigation of sample material of human origin
- Used to demonstrate the differentiation of squamous cells
Specifications
Specifications
| Packaging | Plastic Bottle |
| Quantity | 1L |
| Storage Requirements | 15° to 25°C |